Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethyl-2-butanol, MP Biomedicals

Catalog No. ICN215959 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN215959 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN215959 Supplier MP Biomedicals, Inc Supplier No. 0521595905

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 594-60-5
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
MDL Number MFCD00004463
InChI Key IKECULIHBUCAKR-UHFFFAOYSA-N
Synonym 2,3-dimethyl-2-butanol, thexyl alcohol, isopropyldimethylcarbinol, 2-butanol, 2,3-dimethyl, acmc-1asco, 2,3-dimethyl-butan-2-ol, 2-butanol,2,3-dimethyl, ikeculihbucakr-uhfffaoysa, ch3 2chc oh ch3 2
PubChem CID 11670
IUPAC Name 2,3-dimethylbutan-2-ol
SMILES CC(C)C(C)(C)O

Specifications

Quantity 10 g
Formula Weight 102.18
Chemical Name or Material 2, 3-Dimethyl-2-butanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.