accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)CH3SS(R)CH3CH3

CAS RN 5943-30-6

IUPAC Name: (2R)-2-[(2S)-butan-2-yldisulfanyl]butane
Synonyms: (2R)-2-[(2S)-butan-2-yldisulfanyl]butane
Molecular Weight (g/mol): 178.35
Molecular Formula: C8H18S2
InChi Key: QTWKINKGAHTPFJ-OCAPTIKFSA-N
SMILES: CC[C@H](C)SS[C@H](C)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results