Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sec-Butyl disulfide, 98%, Thermo Scientific™

Catalog No. AC294610250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC294610250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC294610250 Supplier Thermo Scientific Chemicals Supplier No. 294610250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5943-30-6
Molecular Formula C8H18S2
Molecular Weight (g/mol) 178.352
InChI Key QTWKINKGAHTPFJ-UHFFFAOYSA-N
Synonym sec-butyl disulfide, di-sec-butyl disulfide, disulfide, bis 1-methylpropyl, di-sec-butyldisulfide, 2-sec-butyldisulfanyl butane, dibutan-2-yl disulfide, disulfide,bis 1-methylpropyl, 2-butan-2-yldisulfanyl butane, sec-butyldisulfide, bis sec-butyl disulphide
PubChem CID 94314
IUPAC Name 2-(butan-2-yldisulfanyl)butane
SMILES CCC(C)SSC(C)CC

Specifications

Density 0.95g/mL
Assay Percent Range 98%
Linear Formula [C2H5CH(CH3)]2S2
Quantity 25 g
Formula Weight 178.35
Chemical Name or Material sec-Butyl disulfide, 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.