accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSOOFFFOSOOFFF

CAS RN 594838-61-6

IUPAC Name: 6-(trifluoromethanesulfonyloxy)anthracen-2-yl trifluoromethanesulfonate
Synonyms: 6-(trifluoromethanesulfonyloxy)anthracen-2-yl trifluoromethanesulfonate
Molecular Weight (g/mol): 474.34
Molecular Formula: C16H8F6O6S2
InChi Key: ZGOORSQUXBKABZ-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)OC1=CC2=CC3=CC=C(OS(=O)(=O)C(F)(F)F)C=C3C=C2C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results