Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anthracene-2,6-diyl Bis(trifluoromethanesulfonate) 98.0+%, TCI America™
SDP

Catalog No. A3127200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A3127200MG 200 mg
A31271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A3127200MG Supplier TCI America Supplier No. A3127200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 594838-61-6
Molecular Formula C16H8F6O6S2
Molecular Weight (g/mol) 474.344
InChI Key ZGOORSQUXBKABZ-UHFFFAOYSA-N
Synonym 2,6-Bis(trifluoromethanesulfonyloxy)anthracene
PubChem CID 57587335
IUPAC Name [6-(trifluoromethylsulfonyloxy)anthracen-2-yl] trifluoromethanesulfonate
SMILES C1=CC(=CC2=CC3=C(C=C21)C=C(C=C3)OS(=O)(=O)C(F)(F)F)OS(=O)(=O)C(F)(F)F

Specifications

Melting Point 118°C
Color White-Yellow
Quantity 200 mg
Formula Weight 474.34
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Anthracene-2,6-diyl Bis(trifluoromethanesulfonate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.