accessibility menu, dialog, popup

UserName

Viewing profile as user:
HONHCH2NHONHCH2NOSOOO

CAS RN 5949-16-6

IUPAC Name: bis(5-ethenyl-2-[hydroxy(quinolin-4-yl)methyl]-1-azabicyclo[2.2.2]octan-1-ium) sulfate
Synonyms: bis(5-ethenyl-2-[hydroxy(quinolin-4-yl)methyl]-1-azabicyclo[2.2.2]octan-1-ium) sulfate
Molecular Weight (g/mol): 686.87
Molecular Formula: C38H46N4O6S
InChi Key: WBBHOISPYYYBTC-UHFFFAOYNA-N
SMILES: [O-]S([O-])(=O)=O.OC(C1CC2CC[NH+]1CC2C=C)C1=C2C=CC=CC2=NC=C1.OC(C1CC2CC[NH+]1CC2C=C)C1=C2C=CC=CC2=NC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results