Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cinchonine Sulfate Dihydrate 98.0+%, TCI America™
SDP

Catalog No. C079125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C079125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C079125G Supplier TCI America Supplier No. C079125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5949-16-6
Molecular Formula C38H46N4O6S
Molecular Weight (g/mol) 686.868
MDL Number MFCD00035639
InChI Key WBBHOISPYYYBTC-VVECRAFTSA-N
PubChem CID 131632545
IUPAC Name (S)-[(2S,4R,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-quinolin-4-ylmethanol;sulfuric acid
SMILES C=CC1CN2CCC1CC2C(C3=CC=NC4=CC=CC=C34)O.C=CC1CN2CCC1CC2C(C3=CC=NC4=CC=CC=C34)O.OS(=O)(=O)O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 686.87
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Cinchonine Sulfate Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.