accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 595-30-2

IUPAC Name: 1,8,8-trimethyl-3-oxabicyclo[3.2.1]octane-2,4-dione
Synonyms: 1,8,8-trimethyl-3-oxabicyclo[3.2.1]octane-2,4-dione
Molecular Weight (g/mol): 182.22
Molecular Formula: C10H14O3
InChi Key: VFZDNKRDYPTSTP-UHFFFAOYNA-N
SMILES: CC1(C)C2CCC1(C)C(=O)OC2=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)