accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNClCl

CAS RN 59514-89-5

IUPAC Name: 2,4-dichloro-1,8-naphthyridine
Synonyms: 2,4-dichloro-1,8-naphthyridine
Molecular Weight (g/mol): 199.03
Molecular Formula: C8H4Cl2N2
InChi Key: CCQHBZIXZFQYSO-UHFFFAOYSA-N
SMILES: ClC1=NC2=NC=CC=C2C(Cl)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results