Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloro-1,8-naphthyridine 96.0+%, TCI America™
SDP

Catalog No. D48961G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48961G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D48961G Supplier TCI America Supplier No. D48961G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 59514-89-5
Molecular Formula C8H4Cl2N2
Molecular Weight (g/mol) 199.034
MDL Number MFCD00234380
InChI Key CCQHBZIXZFQYSO-UHFFFAOYSA-N
Synonym 2,4-Dichloro-1,8-diazanaphthalene, 2,4-Dichloro-pyrido[2,3-b]pyridine
PubChem CID 18449302
IUPAC Name 2,4-dichloro-1,8-naphthyridine
SMILES C1=CC2=C(N=C1)N=C(C=C2Cl)Cl

Specifications

Melting Point 125°C
Color Yellow
Quantity 1 g
Formula Weight 199.03
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichloro-1,8-naphthyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.