accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 59528-27-7

IUPAC Name: 1-(4-iodophenyl)methanamine hydrochloride
Synonyms: 1-(4-iodophenyl)methanamine hydrochloride
Molecular Weight (g/mol): 269.51
Molecular Formula: C7H9ClIN
InChi Key: GBJMURRFWZREHE-UHFFFAOYSA-N
SMILES: Cl.NCC1=CC=C(I)C=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results