accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3

CAS RN 59558-23-5

IUPAC Name: 4-methylphenyl octanoate
Synonyms: P-cresyl octanoate
Molecular Weight (g/mol): 234.34
Molecular Formula: C15H22O2
InChi Key: ALRYNTSLFYRKGF-UHFFFAOYSA-N
SMILES: CCCCCCCC(=O)OC1=CC=C(C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results