Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Tolyl n-Octanoate 96.0+%, TCI America™
SDP

Catalog No. C114725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C114725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C114725ML Supplier TCI America Supplier No. C114725ML

Chemical Identifiers

CAS 59558-23-5
Molecular Formula C15H22O2
Molecular Weight (g/mol) 234.339
MDL Number MFCD00027318
InChI Key ALRYNTSLFYRKGF-UHFFFAOYSA-N
Synonym n-Octanoic Acid p-Tolyl Ester
PubChem CID 43046
IUPAC Name (4-methylphenyl) octanoate
SMILES CCCCCCCC(=O)OC1=CC=C(C=C1)C

Specifications

Color Yellow
Boiling Point 175°C
Quantity 25 mL
Formula Weight 234.34
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material p-Tolyl n-Octanoate
RTECSNumber RH0787500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.