accessibility menu, dialog, popup

UserName

Viewing profile as user:
ICH3NH3CH3CHO

CAS RN 5957-17-5

IUPAC Name: triethyl(2-hydroxyethyl)azanium iodide
Synonyms: triethylcholine
Molecular Weight (g/mol): 273.16
Molecular Formula: C8H20INO
InChi Key: XQDHXDORJFXNDX-UHFFFAOYSA-M
SMILES: [I-].CC[N+](CC)(CC)CCO
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results