Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Hydroxyethyl)triethylammonium Iodide 98.0+%, TCI America™
SDP

Catalog No. H040710G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H040710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H040710G Supplier TCI America Supplier No. H040710G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5957-17-5
Molecular Formula C8H20INO
Molecular Weight (g/mol) 273.158
MDL Number MFCD00031691
InChI Key XQDHXDORJFXNDX-UHFFFAOYSA-M
Synonym Triethylcholine Iodide
PubChem CID 80057
IUPAC Name triethyl(2-hydroxyethyl)azanium;iodide
SMILES CC[N+](CC)(CC)CCO.[I-]

Specifications

Color White-Yellow
Quantity 10 g
Formula Weight 273.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (2-Hydroxyethyl)triethylammonium Iodide
RTECSNumber BR1215950
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.