accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrCH3NH3COOHO

CAS RN 596-51-0

IUPAC Name: 3-[(2-cyclopentyl-2-hydroxy-2-phenylacetyl)oxy]-1,1-dimethylpyrrolidin-1-ium bromide
Synonyms: glycopyrronium bromide
Molecular Weight (g/mol): 398.34
Molecular Formula: C19H28BrNO3
InChi Key: VPNYRYCIDCJBOM-UHFFFAOYNA-M
SMILES: [Br-].C[N+]1(C)CCC(C1)OC(=O)C(O)(C1CCCC1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (4)
Physical Form 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (4)
  • (4)
  • (1)
  • (2)
  • (4)
  • (1)
  • (4)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)