accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOHO

CAS RN 59604-96-5

IUPAC Name: methyl 2-hydroxy-5,6,7,8-tetrahydronaphthalene-1-carboxylate
Synonyms: methyl 2-hydroxy-5,6,7,8-tetrahydronaphthalene-1-carboxylate
Molecular Weight (g/mol): 206.24
Molecular Formula: C12H14O3
InChi Key: FGZPUQMMDVJQQN-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(O)C=CC2=C1CCCC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results