CAS RN 59626-33-4
IUPAC Name:
2-tribromomethanesulfonylpyridine
Synonyms:
2-tribromomethanesulfonylpyridine
Molecular Weight (g/mol):
393.88
Molecular Formula:
C6H4Br3NO2S
InChi Key:
FRCQMXHPNJVPJC-UHFFFAOYSA-N
SMILES:
BrC(Br)(Br)S(=O)(=O)C1=CC=CC=N1
1
–
1
of
1
results
1
2-Pyridyl Tribromomethyl Sulfone 98.0+%, TCI America™
CAS: 59626-33-4 Molecular Formula: C6H4Br3NO2S Molecular Weight (g/mol): 393.875 MDL Number: MFCD02093493 InChI Key: FRCQMXHPNJVPJC-UHFFFAOYSA-N Synonym: 2-(Tribromomethanesulfonyl)pyridine PubChem CID: 11315382 IUPAC Name: 2-(tribromomethylsulfonyl)pyridine SMILES: C1=CC=NC(=C1)S(=O)(=O)C(Br)(Br)Br