Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Pyridyl Tribromomethyl Sulfone 98.0+%, TCI America™
SDP

Catalog No. P13775G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13775G 5 g
P137725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13775G Supplier TCI America Supplier No. P13775G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 59626-33-4
Molecular Formula C6H4Br3NO2S
Molecular Weight (g/mol) 393.875
MDL Number MFCD02093493
InChI Key FRCQMXHPNJVPJC-UHFFFAOYSA-N
Synonym 2-(Tribromomethanesulfonyl)pyridine
PubChem CID 11315382
IUPAC Name 2-(tribromomethylsulfonyl)pyridine
SMILES C1=CC=NC(=C1)S(=O)(=O)C(Br)(Br)Br

Specifications

Color White
Quantity 5 g
Formula Weight 393.88
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Pyridyl Tribromomethyl Sulfone
EINECSNumber (5)-0729
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.