CAS RN 5964-35-2
IUPAC Name:
tetrapotassium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
Synonyms:
tetrapotassium ion(4-) edta
Molecular Weight (g/mol):
444.61
Molecular Formula:
C10H12K4N2O8
InChi Key:
JZBRFIUYUGTUGG-UHFFFAOYSA-J
SMILES:
[K+].[K+].[K+].[K+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
1
–
1
of
1
results
1
MP Biomedicals, Inc EDTA, Tetrapotassium Salt, 45% Soln, MP Biomedicals
CAS: 5964-35-2 Molecular Formula: C10H12K4N2O8 Molecular Weight (g/mol): 444.605 InChI Key: JZBRFIUYUGTUGG-UHFFFAOYSA-J Synonym: tetrapotassium edta,edta tetrapotassium salt,potassium ethylenediaminetetraacetate,unii-4ttl387kw9,tetrapotassium ethylenediaminetetraacetate,ethylenediaminetetraacetic acid tetrapotassium salt,ethylenediaminetetraacetic acid potassium salt,tetrapotassium ion 4-edta,60-00-4 parent,n,n'-ethylenebis n-carboxymethyl aminoacetic acid, potassium salt PubChem CID: 62595 IUPAC Name: tetrapotassium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate SMILES: C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].[K+].[K+].[K+].[K+]