Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDTA, Tetrapotassium Salt, 45% Soln, MP Biomedicals

Catalog No. ICN210859 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN210859 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN210859 Supplier MP Biomedicals, Inc Supplier No. 0521085980

Chemical Identifiers

CAS 5964-35-2
Molecular Formula C10H12K4N2O8
Molecular Weight (g/mol) 444.605
InChI Key JZBRFIUYUGTUGG-UHFFFAOYSA-J
Synonym tetrapotassium edta, edta tetrapotassium salt, potassium ethylenediaminetetraacetate, unii-4ttl387kw9, tetrapotassium ethylenediaminetetraacetate, ethylenediaminetetraacetic acid tetrapotassium salt, ethylenediaminetetraacetic acid potassium salt, tetrapotassium ion 4-edta, 60-00-4 parent, n,n'-ethylenebis n-carboxymethyl aminoacetic acid, potassium salt
PubChem CID 62595
IUPAC Name tetrapotassium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate
SMILES C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].[K+].[K+].[K+].[K+]

Specifications

Formula Weight 452.23
Quantity 100 g
Chemical Name or Material EDTA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.