accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5965-53-7

H3COOOOOCH3

CAS RN 5965-53-7

IUPAC Name: 1,5-diethyl 2-oxopentanedioate
Synonyms: 1,5-diethyl 2-oxopentanedioate
Molecular Weight (g/mol): 202.21
Molecular Formula: C9H14O5
InChi Key: NGRAIMFUWGFAEM-UHFFFAOYSA-N
SMILES: CCOC(=O)CCC(=O)C(=O)OCC
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results