Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl oxalpropionate, Thermo Scientific™

Catalog No. AC160141000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC160141000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC160141000 Supplier Thermo Scientific Chemicals Supplier No. 160141000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5965-53-7
Molecular Formula C9H14O5
Molecular Weight (g/mol) 202.206
InChI Key NGRAIMFUWGFAEM-UHFFFAOYSA-N
Synonym diethyl 2-oxoglutarate, diethyl alpha-ketoglutarate, unii-d9h7kzf90h, diethyl 2-oxo-glutarate, d9h7kzf90h, 1,5-diethyl 2-oxopentanedioate, diethyl 2-oxopentane-1,5-dicarboxylate, zlchem 727, diethyl oxalpropanoate, .alpha.-ketoglutaric acid diethyl ester
PubChem CID 80070
IUPAC Name diethyl 2-oxopentanedioate
SMILES CCOC(=O)CCC(=O)C(=O)OCC

Specifications

Density 1.07g/mL
Assay Percent Range 97%
Linear Formula C2H5O2CCH(CH3)COCO2C2H5
Quantity 100 g
Beilstein 03, 794
Merck Index 15, 5350
Formula Weight 202.21
Chemical Name or Material Diethyl oxalpropionate, 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.