accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 59662-26-9

H3CO

CAS RN 59662-26-9

IUPAC Name: 1-{[1,1'-biphenyl]-4-yl}hexan-1-one
Synonyms: 1-{[1,1'-biphenyl]-4-yl}hexan-1-one
Molecular Weight (g/mol): 252.36
Molecular Formula: C18H20O
InChi Key: HHVQDDCTUWJMFP-UHFFFAOYSA-N
SMILES: CCCCCC(=O)C1=CC=C(C=C1)C1=CC=CC=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results