Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hexanoylbiphenyl, TCI America™
SDP

Catalog No. H071010G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H071010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H071010G Supplier TCI America Supplier No. H071010G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 59662-26-9
Molecular Formula C18H20O
Molecular Weight (g/mol) 252.357
MDL Number MFCD00143349
InChI Key HHVQDDCTUWJMFP-UHFFFAOYSA-N
Synonym 4-Caproylbiphenyl, 4-Hexanoyldiphenyl
PubChem CID 602492
IUPAC Name 1-(4-phenylphenyl)hexan-1-one
SMILES CCCCCC(=O)C1=CC=C(C=C1)C2=CC=CC=C2

Specifications

Melting Point 96°C
Color White
Boiling Point 194°C
Quantity 10 g
Formula Weight 252.36
Physical Form Crystalline Powder
Chemical Name or Material 4-Hexanoylbiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.