accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 59662-31-6

IUPAC Name: 4-hexyl-1,1'-biphenyl
Synonyms: 4-hexyl-1,1'-biphenyl
Molecular Weight (g/mol): 238.37
Molecular Formula: C18H22
InChi Key: XRPVQXPWEVJKTN-UHFFFAOYSA-N
SMILES: CCCCCCC1=CC=C(C=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results