Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hexylbiphenyl 99%, Thermo Scientific™

Catalog No. AC206590050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC206590050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC206590050 Supplier Thermo Scientific Chemicals Supplier No. 206590050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59662-31-6
Molecular Formula C18H22
Molecular Weight (g/mol) 238.37
MDL Number MFCD00009527
InChI Key XRPVQXPWEVJKTN-UHFFFAOYSA-N
Synonym 4-hexylbiphenyl, 4-hexyl-1,1'-biphenyl, 4-n-hexylbiphenyl, 1-hexyl-4-phenyl-benzene, acmc-20amhe, pubchem2517, 4-hexyl-1-phenylbenzene, maybridge1_001959, 1,1'-biphenyl, 4-hexyl
PubChem CID 108797
IUPAC Name 4-hexyl-1,1'-biphenyl
SMILES CCCCCCC1=CC=C(C=C1)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Density 0.98
Infrared Spectrum Authentic
Linear Formula CH3(CH2)5C6H4C6H5
Quantity 5 g
Beilstein 05, IV, 1987
Formula Weight 238.37
Percent Purity ≥98.5%
Physical Form Adhering Chunks and Flakes
Chemical Name or Material 4-Hexylbiphenyl, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.