accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHO

CAS RN 59662-48-5

IUPAC Name: 4'-hexyl-[1,1'-bi(cyclohexane)]-4-carboxylate
Synonyms: 4'-hexyl-[1,1'-bi(cyclohexane)]-4-carboxylate
Molecular Weight (g/mol): 293.47
Molecular Formula: C19H33O2
InChi Key: NWHAWRQJZXUDLX-UHFFFAOYSA-M
SMILES: CCCCCCC1CCC(CC1)C1CCC(CC1)C([O-])=O
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)