Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Hexylphenyl)benzoic Acid 96.0+%, TCI America™
SDP

Catalog No. H07115G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H07115G Supplier TCI America Supplier No. H07115G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 59662-48-5
Molecular Formula C19H22O2
Molecular Weight (g/mol) 282.38
MDL Number MFCD00143232
InChI Key ROJCBWVSXWIGAL-UHFFFAOYSA-N
Synonym 4-4-hexylphenyl benzoic acid, 4-hexyl-4'-biphenylcarboxylic acid, 4-hexyl-4'-carboxybiphenyl, 4-n-hexylbiphenyl-4'-carboxylic acid, 4'-hexyl-1,1'-biphenyl-4-carboxylic acid, pubchem9069, acmc-209mez, 4'-hexylbiphenyl-4-carboxylic acid, 4-hexylbiphenyl-4'-carboxylic acid, 4'-hexyl-biphenyl-4-carboxylic acid
PubChem CID 13554588
IUPAC Name 4'-hexyl-[1,1'-biphenyl]-4-carboxylic acid
SMILES CCCCCCC1=CC=C(C=C1)C1=CC=C(C=C1)C(O)=O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 282.38
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Hexylphenyl)benzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.