accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2ONaNaOOO

CAS RN 5968-11-6

IUPAC Name: disodium hydrate carbonate
Synonyms: disodium hydrate carbonate
Molecular Weight (g/mol): 124
Molecular Formula: CH2Na2O4
InChi Key: MQRJBSHKWOFOGF-UHFFFAOYSA-L
SMILES: O.[Na+].[Na+].[O-]C([O-])=O
Molecular Weight (g/mol) 
  • (24)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (7)
Grade 
  • (1)
  • (5)
  • (6)
  • (1)
  • (3)
  • (3)
  • (4)
  • (2)
Percent Purity 
  • (7)
  • (2)
  • (7)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)
  • (4)
  • (3)
  • (2)
  • (4)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (13)
  • (11)
  • (2)
  • (2)
  • (1)
  • (3)
  • (11)
  • (7)
  • (1)
  • (7)
  • (1)
  • (1)
  • (24)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (7)
  • (1)
  • (5)
  • (6)
  • (1)
  • (3)
  • (3)
  • (4)
  • (2)
  • (7)
  • (2)
  • (7)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (3)
  • (2)
  • (4)
  • (3)
  • (2)