accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOOOOOHNOSNHOHN

CAS RN 596820-83-6

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 3-{2-[2-(5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanamido)ethoxy]ethoxy}propanoate
Synonyms: 2,5-dioxopyrrolidin-1-yl 3-{2-[2-(5-{2-oxo-hexahydrothieno[3,4-d]imidazol-4-yl}pentanamido)ethoxy]ethoxy}propanoate
Molecular Weight (g/mol): 500.57
Molecular Formula: C21H32N4O8S
InChi Key: UCJSJBIJPAZUGU-UHFFFAOYNA-N
SMILES: O=C(CCCCC1SCC2NC(=O)NC12)NCCOCCOCCC(=O)ON1C(=O)CCC1=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results