Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Succinimidyl 9-(Biotinamido)-4,7-dioxanonanoate, TCI America™
SDP

Catalog No. S095525MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S095525MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S095525MG Supplier TCI America Supplier No. S095525MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 596820-83-6
Molecular Formula C21H32N4O8S
Molecular Weight (g/mol) 500.567
InChI Key UCJSJBIJPAZUGU-AVYPCKFXSA-N
Synonym 9-(Biotinamido)-4,7-dioxanonanoic Acid N-Succinimidyl Ester, Biotin-PEG2-NHS
PubChem CID 91757819
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 3-[2-[2-[5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoylamino]ethoxy]ethoxy]propanoate
SMILES C1CC(=O)N(C1=O)OC(=O)CCOCCOCCNC(=O)CCCCC2C3C(CS2)NC(=O)N3

Specifications

Color White
Quantity 25 mg
Formula Weight 500.57
Physical Form Crystalline Powder
Chemical Name or Material N-Succinimidyl 9-(Biotinamido)-4,7-dioxanonanoate
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.