accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHHO

CAS RN 59719-74-3

IUPAC Name: cyclopentane-1,3-diol
Synonyms: 1,3-cyclopentanediol, cis-
Molecular Weight (g/mol): 102.13
Molecular Formula: C5H10O2
InChi Key: NUUPJBRGQCEZSI-UHFFFAOYNA-N
SMILES: OC1CCC(O)C1
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)