Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Cyclopentanediol (cis- and trans- mixture) 95.0+%, TCI America™
SDP

Catalog No. C25341G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C25341G Supplier TCI America Supplier No. C25341G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 59719-74-3
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00001367
InChI Key NUUPJBRGQCEZSI-UHFFFAOYSA-N
Synonym 1,3-cyclopentanediol, 1,3-cyclopentanediol, cis, 1,3-cyclopentanediol, mixture of cis and trans, 1,3-cyclopentandiol, 1,4-cyclopentanediol, cyclopentan-1,3-diol, 3-hydroxy cylcopentanol, cyclopentane 1,3-diol, acmc-20ap4n, 1,3-dihydroxycyclopentane
PubChem CID 100165
IUPAC Name cyclopentane-1,3-diol
SMILES C1CC(CC1O)O

Specifications

Color Yellow
Boiling Point 91°C
Quantity 1 g
Formula Weight 102.13
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Cyclopentanediol (cis- and trans- mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.