accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3CH3OOHNOOH3COHO

CAS RN 59768-74-0

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-4-methoxy-4-oxobutanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-4-methoxy-4-oxobutanoic acid
Molecular Weight (g/mol): 247.25
Molecular Formula: C10H17NO6
InChi Key: WFPSMPYVXFVVFA-UHFFFAOYNA-N
SMILES: COC(=O)CC(NC(=O)OC(C)(C)C)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results