Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(+)-2-Octanol

Catalog No. 1479825G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-798-25G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 14-798-25G Supplier Sigma Aldrich Supplier No. 1479825G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-(+)-2-Octanol can be used:To prepare the solution of 5-(benzyloxy)-isophthalic acid derivative, which is used as a 2D chiral self-assembly system comprising pyrolytic graphite.As a chiral template in the study of enantioselective glycidol esterification.As a starting material for the preparation of (+)-(S)-2-octyI tosylate, an intermediate used to prepare (-)-™-2-halo and azido octanes.

Specifications

CAS 59768-74-0
Molecular Formula C10H17NO6
Linear Formula CH3O2CCH2CH[NHCO2C(CH3)3]CO2H
Refractive Index n20/D 1.426
MDL Number MFCD00078971
Quantity 5 g
Synonym Boc-L-aspartic acid 4-methyl ester
Molecular Weight (g/mol) 247.25 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.