accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOH3C

CAS RN 598-04-9

IUPAC Name: 1-(butane-1-sulfonyl)butane
Synonyms: butane, 1,1'-sulfonylbis-
Molecular Weight (g/mol): 178.29
Molecular Formula: C8H18O2S
InChi Key: AIDFJGKWTOULTC-UHFFFAOYSA-N
SMILES: CCCCS(=O)(=O)CCCC
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)