Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibutyl Sulfone 97.0+%, TCI America™
SDP

Catalog No. D117225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D117225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D117225G Supplier TCI America Supplier No. D117225G

Chemical Identifiers

CAS 598-04-9
Molecular Formula C8H18O2S
Molecular Weight (g/mol) 178.29
MDL Number MFCD00007571
InChI Key AIDFJGKWTOULTC-UHFFFAOYSA-N
Synonym dibutyl sulfone, butyl sulfone, butane, 1,1'-sulfonylbis, di-n-butyl sulfone, dibutyl sulphone, n-butyl sulfone, 1-butylsulfonyl butane, butylsulfonyl butane, dibutylsulfone, di-n-butylsulfone
PubChem CID 69014
IUPAC Name 1-butylsulfonylbutane
SMILES CCCCS(=O)(=O)CCCC

Specifications

Melting Point 46°C
Color White
Quantity 25 g
Formula Weight 178.29
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dibutyl Sulfone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.