accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH2HOBr

CAS RN 598-19-6

IUPAC Name: 2-bromoprop-2-en-1-ol
Synonyms: 2-bromo-2-propene-1-ol C3H5BrO
Molecular Weight (g/mol): 136.98
Molecular Formula: C3H5BrO
InChi Key: MDFFZNIQPLKQSG-UHFFFAOYSA-N
SMILES: OCC(Br)=C
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)