Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromoallyl Alcohol 97.0+%, TCI America™
SDP

Catalog No. B53475G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B53475G 5 g
B53471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B53475G Supplier TCI America Supplier No. B53475G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 598-19-6
Molecular Formula C3H5BrO
Molecular Weight (g/mol) 136.976
MDL Number MFCD09265077
InChI Key MDFFZNIQPLKQSG-UHFFFAOYSA-N
Synonym 2-Bromo-2-propen-1-ol
PubChem CID 446627
ChEBI CHEBI:49514
IUPAC Name 2-bromoprop-2-en-1-ol
SMILES C=C(CO)Br

Specifications

Boiling Point 150°C
Quantity 5 g
Formula Weight 136.98
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromoallyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.