accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3HC

CAS RN 598-23-2

IUPAC Name: 3-methylbut-1-yne
Synonyms: 1-butyne, 3-methyl-
Molecular Weight (g/mol): 68.12
Molecular Formula: C5H8
InChi Key: USCSRAJGJYMJFZ-UHFFFAOYSA-N
SMILES: CC(C)C#C
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)