Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Methyl-1-butyne

Catalog No. 7457235G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-572-35G 5 g
74-572-325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 74-572-35G Supplier Sigma Aldrich Supplier No. 7457235G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-methyl-1-butyne is a terminal alkyne that can be used to prepare:
  • Maleimide-fused cyclopentenones by reacting with n-substituted maleimides via co2(co)8-mediated pausonkhand reaction
  • An alkyne carboxylic acid (alkyne segment c) intermediate, which is used in the total synthesis of oscillatoxin d and its analogs

Specifications

CAS 598-23-2
Density 0.666 g/mL (at 25°C (literature))
Molecular Formula C5H8
Refractive Index n20/D 1.374
Linear Formula C5H8
MDL Number MFCD00039853
Quantity 5 g
Synonym 1-Isopentyne; 2-Methyl-3-butyne; 3,3-Dimethyl-1-propyne; Isopropylacetylene; Isopropylethyne
Molecular Weight (g/mol) 68.12
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.