accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5982-56-9

IUPAC Name: ethane-1,2-disulfonic acid dihydrate
Synonyms: ethanedisulfonic acid dihydrate
Molecular Weight (g/mol): 226.21
Molecular Formula: C2H10O8S2
InChi Key: DSGUSEBCDAKBCM-UHFFFAOYSA-N
SMILES: O.O.OS(=O)(=O)CCS(O)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results