accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 59855-11-7

IUPAC Name: [amino(4-carbamoylphenyl)methylidene]azanium
Synonyms: [amino(4-carbamoylphenyl)methylidene]azanium
Molecular Weight (g/mol): 164.19
Molecular Formula: C8H10N3O
InChi Key: HKMIWYWBIYCXFQ-UHFFFAOYSA-O
SMILES: NC(=[NH2+])C1=CC=C(C=C1)C(N)=O
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)