Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amidinobenzamide hydrochloride, 99%, Thermo Scientific™

Catalog No. AC224400100
Change view
Click to view available options
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC224400100 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC224400100 Supplier Thermo Scientific Chemicals Supplier No. 224400100

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 59855-11-7
CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C8H10ClN3O
Linear Formula H2NC(NH)C6H4CONH2·HCl
Quantity 10g
Beilstein 09, III, 4255
Synonym 4-amidinobenzamide hydrochloride, 4-carbamimidoylbenzamide hydrochloride, 4-amidino-benzoic acid amide hydrochloride, acmc-1awl9, 4-amidinobenzamide hcl, 4-carbamimidoyl-benzamide,hydrochloride, 4-amidinobenzoic acid amide hydrochloride, 4-amino imino methyl benzamide hydrochloride, benzamide, 4-aminoiminomethyl-, hydrochloride, benzamide,4-aminoiminomethyl-, hydrochloride 1:1
InChI Key IYIAWOSYBLPUNL-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1C(=N)N)C(=O)N.Cl
IUPAC Name 4-carbamimidoylbenzamide;hydrochloride
Molecular Weight (g/mol) 199.638
PubChem CID 2724293
Formula Weight 199.64
Percent Purity ≥98.5% (on dry substance)
Chemical Name or Material 4-Amidinobenzamide hydrochloride, 99%
Show More Show Less
Hazard Statement GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Wash face, hands and any exposed skin thoroughly after handling. Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.