CAS RN 59857-86-2
IUPAC Name:
methyl 1-methyl-5-oxopyrrolidine-3-carboxylate
Synonyms:
methyl 1-methyl-5-oxopyrrolidine-3-carboxylate
Molecular Weight (g/mol):
157.17
Molecular Formula:
C7H11NO3
InChi Key:
JFZRRXGXEHUJCI-UHFFFAOYNA-N
SMILES:
COC(=O)C1CN(C)C(=O)C1
1
–
1
of
1
results
1
Methyl 1-methyl-5-oxopyrrolidine-3-carboxylate, 97%, Thermo Scientific™
CAS: 59857-86-2 Molecular Formula: C7H11NO3 Molecular Weight (g/mol): 157.17 MDL Number: MFCD00015898 InChI Key: JFZRRXGXEHUJCI-UHFFFAOYNA-N PubChem CID: 108402 IUPAC Name: methyl 1-methyl-5-oxopyrrolidine-3-carboxylate SMILES: COC(=O)C1CN(C)C(=O)C1