accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNNNN

CAS RN 5987-76-8

IUPAC Name: 7-methyl-5-phenyl-[1,2,3,4]tetrazolo[1,5-a]quinazoline
Synonyms: 7-methyl-5-phenyl-[1,2,3,4]tetrazolo[1,5-a]quinazoline
Molecular Weight (g/mol): 261.29
Molecular Formula: C15H11N5
InChi Key: HBIRBQGJJVJKAQ-UHFFFAOYSA-N
SMILES: CC1=CC=C2N3N=NN=C3N=C(C3=CC=CC=C3)C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results