Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',3',5'-Tri-O-acetyl-6-chloro-2-iodopurine Riboside 97.0+%, TCI America™
SDP

Catalog No. T26915G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26915G 5 g
T26911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26915G Supplier TCI America Supplier No. T26915G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5987-76-8
Molecular Formula C16H16ClIN4O7
Molecular Weight (g/mol) 538.679
MDL Number MFCD03701153
InChI Key BYOJIJDSBSWKKE-CKRXIKOQSA-N
Synonym 6-Chloro-2-iodo-9-(2,3,5-tri-O-acetyl-beta-D-ribofuranosyl)purine
PubChem CID 46780985
IUPAC Name [(2R,3S,4S,5R)-3,4-diacetyloxy-5-(6-chloro-2-iodopurin-9-yl)oxolan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(O1)N2C=NC3=C2N=C(N=C3Cl)I)OC(=O)C)OC(=O)C

Specifications

Melting Point 184°C
Color Yellow
Quantity 5 g
Formula Weight 538.68
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2′,3′,5′-Tri-O-acetyl-6-chloro-2-iodopurine Riboside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.