accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3HOOHOHHOOOHO

CAS RN 5988-51-2

IUPAC Name: 2,3-dihydroxybutanedioic acid; 2-(dimethylamino)ethan-1-ol
Synonyms: (.+-.)-tartaric acid; dimethylaminoethanol
Molecular Weight (g/mol): 239.22
Molecular Formula: C8H17NO7
InChi Key: UIEGYKVRCKDVKQ-UHFFFAOYNA-N
SMILES: CN(C)CCO.OC(C(O)C(O)=O)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)