Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Dimethylamino)ethanol Hydrogen L-(+)-Tartrate, TCI America™
SDP

Catalog No. D117425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D117425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D117425G Supplier TCI America Supplier No. D117425G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5988-51-2
Molecular Formula C8H17NO7
Molecular Weight (g/mol) 239.22
MDL Number MFCD00066567
InChI Key UIEGYKVRCKDVKQ-UHFFFAOYNA-N
Synonym 2-(Dimethylamino)ethanol L-(+)-Bitartrate
PubChem CID 16212495
IUPAC Name 2,3-dihydroxybutanedioic acid; 2-(dimethylamino)ethan-1-ol
SMILES CN(C)CCO.OC(C(O)C(O)=O)C(O)=O

Specifications

Melting Point 113°C
Color White-Yellow
Quantity 25 g
Formula Weight 239.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Dimethylamino)ethanol Hydrogen L-(+)-Tartrate
RTECSNumber KK7175000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.